accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3ONCH3CH3OOHOH(E/Z)

CAS RN 68842-66-0

IUPAC Name: 3-(diethylamino)-6-{3-[4-(diethylamino)-2-hydroxyphenyl]-2-hydroxy-4-oxocyclobut-2-en-1-ylidene}cyclohexa-2,4-dien-1-one
Synonyms: 3-(diethylamino)-6-{3-[4-(diethylamino)-2-hydroxyphenyl]-2-hydroxy-4-oxocyclobut-2-en-1-ylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 408.5
Molecular Formula: C24H28N2O4
InChi Key: ODWFNEITXILTAS-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC(O)=C(C=C1)C1=C(O)C(C1=O)=C1C=CC(=CC1=O)N(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results