Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine 98.0+%, TCI America™
SDP

Catalog No. B43425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43425G 5 g
B43421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43425G Supplier TCI America Supplier No. B43425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68842-66-0
Molecular Formula C24H28N2O4
Molecular Weight (g/mol) 408.498
MDL Number MFCD00142492
InChI Key ODWFNEITXILTAS-RELWKKBWSA-N
PubChem CID 18806793
IUPAC Name (6E)-3-(diethylamino)-6-[3-[4-(diethylamino)-2-hydroxyphenyl]-2-hydroxy-4-oxocyclobut-2-en-1-ylidene]cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=C(C=C1)C2=C(C(=C3C=CC(=CC3=O)N(CC)CC)C2=O)O)O

Specifications

Quantity 5 g
Formula Weight 408.50
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.