accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNH2NH2HCl(E/Z)

CAS RN 68936-13-0

IUPAC Name: 4-[2-(4-methoxyphenyl)diazen-1-yl]benzene-1,3-diamine hydrochloride
Synonyms: 4-[2-(4-methoxyphenyl)diazen-1-yl]benzene-1,3-diamine hydrochloride
Molecular Weight (g/mol): 278.74
Molecular Formula: C13H15ClN4O
InChi Key: NKGWQPAKARFOPX-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(C=C1)N=NC1=C(N)C=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results