Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxy Red 85.0+%, TCI America™
SDP

Catalog No. M012925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M012925G Supplier TCI America Supplier No. M012925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68936-13-0
Molecular Formula C13H15ClN4O
Molecular Weight (g/mol) 278.74
MDL Number MFCD00060224
InChI Key NKGWQPAKARFOPX-UHFFFAOYSA-N
Synonym 4-Methoxychrysoidine Hydrochloride, 4-(4-Methoxyphenylazo)-1,3-phenylenediamine Monohydrochloride
PubChem CID 44630364
IUPAC Name 4-[(4-methoxyphenyl)diazenyl]benzene-1,3-diamine;hydrochloride
SMILES COC1=CC=C(C=C1)N=NC2=C(C=C(C=C2)N)N.Cl

Specifications

Color Blue
Quantity 25 g
Formula Weight 278.74
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methoxy Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.