CAS RN 68955-22-6
IUPAC Name:
titanium(2+) butan-1-ol propan-2-ol
Synonyms:
titanium(2+) butanol isopropyl alcohol
Molecular Weight (g/mol):
182.09
Molecular Formula:
C7H18O2Ti
InChi Key:
RNEMAYHGZUGSHP-UHFFFAOYSA-N
SMILES:
[Ti++].CC(C)O.CCCCO
1
–
1
of
1
results
1
Titanium(IV) butoxy isopropoxy complexes, VERTEC™ BIP, Thermo Scientific™
CAS: 68955-22-6 Molecular Formula: C7H18O2Ti Molecular Weight (g/mol): 182.09 MDL Number: MFCD01324803 InChI Key: RNEMAYHGZUGSHP-UHFFFAOYSA-N IUPAC Name: titanium(2+) butan-1-ol propan-2-ol SMILES: [Ti++].CC(C)O.CCCCO