Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) butoxy isopropoxy complexes, VERTEC™ BIP, Thermo Scientific™

Catalog No. AA44681A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44681A3 2 kg
AA4468122 100 g
AA4468136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA44681A3 Supplier Thermo Scientific Chemicals Supplier No. 44681A3

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68955-22-6
Molecular Formula C7H18O2Ti
Molecular Weight (g/mol) 182.09
MDL Number MFCD01324803
InChI Key RNEMAYHGZUGSHP-UHFFFAOYSA-N
IUPAC Name titanium(2+) butan-1-ol propan-2-ol
SMILES [Ti++].CC(C)O.CCCCO

Specifications

Physical Form Liquid
Quantity 2 kg
UN Number UN1993
Chemical Name or Material Titanium (IV) butoxy isopropoxy complexes, VERTEC™ BIP
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.