accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOClH3CSOOClCH3

CAS RN 68985-08-0

IUPAC Name: 2,4,6-trimethylbenzene-1,3-disulfonyl dichloride
Synonyms: 2,4,6-trimethylbenzene-1,3-disulfonyl dichloride
Molecular Weight (g/mol): 317.2
Molecular Formula: C9H10Cl2O4S2
InChi Key: LNXKRGBQKLXFTB-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1S(Cl)(=O)=O)S(Cl)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results