accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClH3CNH3CH3CNHClNOH3C

CAS RN 69-05-6

IUPAC Name: dihydrogen 6-chloro-N-[5-(diethylamino)pentan-2-yl]-2-methoxyacridin-9-amine dichloride
Synonyms: dihydrogen quinacrine dichloride
Molecular Weight (g/mol): 472.88
Molecular Formula: C23H32Cl3N3O
InChi Key: UDKVBVICMUEIKS-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCN(CC)CCCC(C)NC1=C2C=C(OC)C=CC2=NC2=CC(Cl)=CC=C12
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results