Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Quinacrine, Dihydrochloride

Catalog No. 551850100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-185-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-185-0100MG Supplier MilliporeSigma™ Supplier No. 551850100MG
Add to Cart
Add to Cart

A non-specific phospholipase A2 (PLA2) inhibitor.

A non-specific phospholipase A2 (PLA2) inhibitor. Acts as an acetylcholine receptor antagonist. Suppresses glibenclamide-sensitive K+-currents (IC50 = 4.4μM). Also inhibits monoamine oxidase (MAO).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins PLA2
CAS 69-05-6
Molecular Formula C23H30CIN3O·2HCl
Formula Weight 472.9g/mol
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.