accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNNSFFF

CAS RN 69-23-8

IUPAC Name: 2-(4-{3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl}piperazin-1-yl)ethan-1-ol
Synonyms: fluphenazine elinol fluorphenazine flufenazin prolixin enanthate fluphenazine (product) prolixine fluphenazinum phthorphenazine vespazine
Molecular Weight (g/mol): 437.53
Molecular Formula: C22H26F3N3OS
InChi Key: PLDUPXSUYLZYBN-UHFFFAOYSA-N
SMILES: OCCN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results