accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3S(R)(R)NHO(R)NH2ON(S)HOO

CAS RN 69-53-4

IUPAC Name: (2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: ampicillin amcill polycillin deripen ampicillin base ampicillin anhydrate tokiocillin amfipen V aminobenzylpenicillin ampicillin anhydrous
Molecular Weight (g/mol): 349.41
Molecular Formula: C16H19N3O4S
InChi Key: AVKUERGKIZMTKX-NJBDSQKTSA-N
SMILES: CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
Color 
  • (5)
  • (2)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Type 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)