accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3CH3S(R)(R)NHOON(S)OO

CAS RN 69-57-8

IUPAC Name: sodium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: penicillin G sodium monosodium benzylpenicillin crystapen sodium penicillin G benzylpenicillinic acid sodium salt sodium benzylpenicillinate benzylpenicillin sodium salt american penicillin benzylpenicillin sodium
Molecular Weight (g/mol): 356.37
Molecular Formula: C16H17N2NaO4S
InChi Key: FCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES: [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Color 
  • (9)
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (1)
Physical Form 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Type 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (9)
  • (2)
  • (6)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (2)
  • (1)
  • (2)