Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G sodium salt, 99.7%,USP, MP Biomedicals™

Catalog No. ICN15606501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15606501 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15606501 Supplier MP Biomedicals, Inc Supplier No. 0215606501

missing translation for 'validMainframeMsg'

Specifications

Type Penicillin G Sodium Salt
Physical Form Powder
CAS 69-57-8
Color White
Quantity 1 μg
Assay Percent Range 0.997
Molecular Formula C16H17N2NaO4S
Synonym Benzylpenicillin
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 356.372
PubChem CID 23668834
ChEBI CHEBI:51765
Formula Weight 356.4
Grade USP
Chemical Name or Material Penicillin G Sodium Salt
Show More Show Less
storageNote1 Protect from light.
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.