CAS RN 69011-15-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
MilliporeSigma™ Supelco™ AmberLite HPR4100 Cl, Chloride Form
Formerly Dowex™ Marathon™ A2 chloride