Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ AmberLite HPR4100 Cl, Chloride Form

Catalog No. 111015193 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5193 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5193 Supplier MilliporeSigma™ Supelco™ Supplier No. 4339341KG
Add to Cart
Add to Cart

Formerly Dowex™ Marathon™ A2 chloride

Dowex Marathon A2 has dimethylethanolamine as functional group.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
Quantity 1 kg
Product Line Marathon
Capacity (Metric) 1.2 meq/mL by wetted bed volume
MDL Number MFCD00211881
CAS 69011-15-0

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.