accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69033-81-4

IUPAC Name: 3-(4-propoxyphenyl)prop-2-enoic acid
Synonyms: 3-(4-propoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: WTYNDSOJMSGRQV-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=CC(O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results