Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxycinnamic Acid 97.0+%, TCI America™
SDP

Catalog No. P189825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P189825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P189825G Supplier TCI America Supplier No. P189825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69033-81-4
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
InChI Key WTYNDSOJMSGRQV-YVMONPNESA-N
Synonym 3-(4-Propoxyphenyl)acrylic Acid
PubChem CID 7316300
IUPAC Name (Z)-3-(4-propoxyphenyl)prop-2-enoic acid
SMILES CCCOC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.