accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690632-12-3

ClNSNH2HCl

CAS RN 690632-12-3

IUPAC Name: 1-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine hydrochloride
Synonyms: 1-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine hydrochloride
Molecular Weight (g/mol): 261.16
Molecular Formula: C10H10Cl2N2S
InChi Key: DBEFCUHDXFEPSJ-UHFFFAOYSA-N
SMILES: Cl.NCC1=CSC(=N1)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results