accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690632-16-7

IUPAC Name: tert-butyl 3-(4-nitrophenoxy)piperidine-1-carboxylate
Synonyms: tert-butyl 3-(4-nitrophenoxy)piperidine-1-carboxylate
Molecular Weight (g/mol): 322.36
Molecular Formula: C16H22N2O5
InChi Key: XPOFZYJUWZHERT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(C1)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results