Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate, Thermo Scientific™

Catalog No. MO07092CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07092CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07092CB Supplier Thermo Scientific Chemicals Supplier No. MO07092CB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 690632-16-7
Molecular Formula C16H22N2O5
Molecular Weight (g/mol) 322.361
MDL Number MFCD05865146
InChI Key XPOFZYJUWZHERT-UHFFFAOYSA-N
Synonym tert-butyl 3-4-nitrophenoxy piperidine-1-carboxylate, tert-butyl 3-4-nitrophenoxy tetrahydro-1 2h-pyridinecarboxylate, tert-butyl-3-4-nitrophenoxy tetrahydro-1 2h-pyridinecarboxylate, 1-piperidinecarboxylicacid, 3-4-nitrophenoxy-, 1,1-dimethylethyl ester, tert-butyl3-4-nitrophenoxy tetrahydro-1 2h-pyridinecarboxylate, 1n-boc 3-4'-nitrophenoxy piperidine, 1n-boc 3-4-nitrophenoxy piperidine, tert-butyl 3-4-nitrophenoxy piperidinecarboxylate, 1-piperidinecarboxylicacid,3-4-nitrophenoxy-,1,1-dimethylethyl ester
PubChem CID 2794781
IUPAC Name tert-butyl 3-(4-nitrophenoxy)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 322.36
Chemical Name or Material Tert-butyl 3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.