accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHN

CAS RN 690632-28-1

IUPAC Name: 1-(piperidine-3-carbonyl)azepane
Synonyms: 1-(piperidine-3-carbonyl)azepane
Molecular Weight (g/mol): 210.32
Molecular Formula: C12H22N2O
InChi Key: RXLCUJOYUJWMPG-UHFFFAOYNA-N
SMILES: O=C(C1CCCNC1)N1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)