Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Azepanyl(3-piperidinyl)methanone, ≥97%, Thermo Scientific™

Catalog No. MO01174DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01174DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01174DA Supplier Thermo Scientific Chemicals Supplier No. MO01174DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 690632-28-1
Molecular Formula C12H22N2O
Molecular Weight (g/mol) 210.321
InChI Key RXLCUJOYUJWMPG-UHFFFAOYSA-N
Synonym 1-azepanyl 3-piperidinyl methanone, 1-piperidin-3-ylcarbonyl azepane, azepan-1-yl piperidin-3-yl methanone, 1-piperidine-3-carbonyl azepane, methanone, hexahydro-1h-azepin-1-yl-3-piperidinyl, 1h-azepine, hexahydro-1-3-piperidinylcarbonyl, azaperhydroepinyl 3-piperidyl ketone, azepan-1-yl-piperidin-3-yl-methanone, 1-3-piperidylcarbonyl azepane, 1-piperid-3-ylcarbonyl azepane
PubChem CID 2794692
IUPAC Name azepan-1-yl(piperidin-3-yl)methanone
SMILES C1CCCN(CC1)C(=O)C2CCCNC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.32
Chemical Name or Material 1-Azepanyl(3-piperidinyl)methanone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.