accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690634-80-1

IUPAC Name: 4-methyl-1-(piperidine-2-carbonyl)piperidine hydrochloride
Synonyms: 4-methyl-1-(piperidine-2-carbonyl)piperidine hydrochloride
Molecular Weight (g/mol): 246.78
Molecular Formula: C12H23ClN2O
InChi Key: ITPHYLQZNJAUDV-UHFFFAOYNA-N
SMILES: Cl.CC1CCN(CC1)C(=O)C1CCCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results