Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methylpiperidino)(2-piperidinyl)methanone hydrochloride, 95%, Thermo Scientific™

Catalog No. MO01177DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01177DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01177DA Supplier Thermo Scientific Chemicals Supplier No. MO01177DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 690634-80-1
Molecular Formula C12H23ClN2O
Molecular Weight (g/mol) 246.779
MDL Number MFCD06407988
InChI Key ITPHYLQZNJAUDV-UHFFFAOYSA-N
Synonym 4-methylpiperidino 2-piperidinyl methanone hydrochloride, 4-methyl-1-piperidinyl 2-piperidinyl methanone hydrochloride, 4-methyl-1-piperidine-2-carbonyl piperidine hydrochloride, 4-methylpiperidin-1-yl piperidin-2-yl methanone hydrochloride, 4-methylpiperidyl 2-piperidyl ketone, chloride, 4-methylpiperidin-1-yl-piperidin-2-ylmethanone hydrochloride, 4-methylpiperidin-1-yl piperidin-2-yl methanone-hydrogen chloride 1/1
PubChem CID 2794697
IUPAC Name (4-methylpiperidin-1-yl)-piperidin-2-ylmethanone;hydrochloride
SMILES CC1CCN(CC1)C(=O)C2CCCCN2.Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 246.78
Percent Purity 95%
Chemical Name or Material (4-Methylpiperidino)(2-piperidinyl)methanone hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.