accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6908-41-4

IUPAC Name: methyl 4-(hydroxymethyl)benzoate
Synonyms: methyl 4-(hydroxymethyl)benzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: VBWFYEFYHJRJER-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CO)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)