Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(Hydroxymethyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. H064025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H064025G 25 g
H06405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H064025G Supplier TCI America Supplier No. H064025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6908-41-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00009984
InChI Key VBWFYEFYHJRJER-UHFFFAOYSA-N
Synonym methyl 4-hydroxymethyl benzoate, benzoic acid, 4-hydroxymethyl-, methyl ester, 4-carbomethoxybenzyl alcohol, 4-hydroxymethyl benzoic acid methyl ester, methyl p-hydroxymethyl benzoate, acmc-209o5w, methyl p-hydroxymethylbenzoate, methyl-4-hydroxymethylbenzoate, p-methoxycarbonylbenzyl alcohol
PubChem CID 81325
IUPAC Name methyl 4-(hydroxymethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CO

Specifications

Melting Point 52°C
Color White
Boiling Point 157°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Hydroxymethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.