CAS RN 691005-38-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ β-Catenin/Tcf Inhibitor V, BC21
The β-Catenin/Tcf Inhibitor V, BC21, also referenced under CAS 691005-38-6, controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Biochemicals applications.