Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Catenin/Tcf Inhibitor V, BC21

Catalog No. 21933410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-933-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-933-410MG Supplier MilliporeSigma™ Supplier No. 21933410MG
Add to Cart
Add to Cart

The β-Catenin/Tcf Inhibitor V, BC21, also referenced under CAS 691005-38-6, controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Biochemicals applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Brown
Description 95% by HPLC
CAS 691005-38-6
Molecular Formula C32H36Cl2Cu2N2O2
Formula Weight 678.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.