accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6911-51-9

IUPAC Name: 2-(thiophen-2-yldisulfanyl)thiophene
Synonyms: disulfide, 2-thienyl
Molecular Weight (g/mol): 230.38
Molecular Formula: C8H6S4
InChi Key: YOLFWWMPGNMXFI-UHFFFAOYSA-N
SMILES: S(SC1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results