Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dithienyl Disulfide 97.0+%, TCI America™
SDP

Catalog No. D37745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37745G 5 g
D37741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37745G Supplier TCI America Supplier No. D37745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6911-51-9
Molecular Formula C8H6S4
Molecular Weight (g/mol) 230.376
MDL Number MFCD00066333
InChI Key YOLFWWMPGNMXFI-UHFFFAOYSA-N
Synonym 2-thienyl disulfide, thiophene, 2,2'-dithiobis, disulfide, 2-thienyl, di-2-thienyl disulfide, 2,2'-dithiodithiophene, di 2-thienyl disulfide, dithienyl disulfide, 2,2'-dithiobis thiophene, 2,2'-bis thienyl disulfide, alpha,alpha'-dithienyl disulfide
PubChem CID 23347
IUPAC Name 2-(thiophen-2-yldisulfanyl)thiophene
SMILES C1=CSC(=C1)SSC2=CC=CS2

Specifications

Melting Point 58°C
Boiling Point 132°C
Quantity 5 g
Formula Weight 230.38
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dithienyl Disulfide
RTECSNumber JO1987000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.