accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6912-12-5

IUPAC Name: hydrogen dimethylamine bromide
Synonyms: hydrogen dimethylamine bromide
Molecular Weight (g/mol): 126
Molecular Formula: C2H8BrN
InChi Key: FFJMLWSZNCJCSZ-UHFFFAOYSA-N
SMILES: [H+].[Br-].CNC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results