Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylamine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. D50925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50925G 5 g
D50921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50925G Supplier TCI America Supplier No. D50925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6912-12-5
Molecular Formula C2H8BrN
Molecular Weight (g/mol) 126.00
MDL Number MFCD00051983
InChI Key FFJMLWSZNCJCSZ-UHFFFAOYSA-N
Synonym Dimethylammonium Bromide
PubChem CID 12199014
IUPAC Name N-methylmethanamine;hydrobromide
SMILES CNC.Br

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 126.00
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylamine Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.