accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONHCl

CAS RN 69135-90-6

IUPAC Name: N-phenylpyridine-3-carboxamide hydrochloride
Synonyms: nicotinanilide hydrochloride
Molecular Weight (g/mol): 234.68
Molecular Formula: C12H11ClN2O
InChi Key: SQFVWDGIESXVGY-UHFFFAOYSA-N
SMILES: Cl.O=C(NC1=CC=CC=C1)C1=CN=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results