Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylnicotinamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P019910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P019910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P019910G Supplier TCI America Supplier No. P019910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69135-90-6
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.683
MDL Number MFCD00060194
InChI Key SQFVWDGIESXVGY-UHFFFAOYSA-N
Synonym Nicotinanilide Hydrochloride
PubChem CID 23296855
IUPAC Name N-phenylpyridine-3-carboxamide;hydrochloride
SMILES C1=CC=C(C=C1)NC(=O)C2=CN=CC=C2.Cl

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 10 g
Formula Weight 234.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylnicotinamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.