accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCHNCH

CAS RN 6921-28-4

IUPAC Name: bis(prop-2-yn-1-yl)amine
Synonyms: di-2-propynylamine
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: RGSODMOUXWISAG-UHFFFAOYSA-N
SMILES: C#CCNCC#C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)