Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dipropargylamine

Catalog No. 7457311G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-573-11G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-573-11G Supplier Sigma Aldrich Supplier No. 7457311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dipropargylamine

Specifications

CAS 6921-28-4
Density 0.900 g/mL (at 25°C)
Molecular Formula C6H7N
Refractive Index n20/D 1.477
Linear Formula C6H7N
MDL Number MFCD00008574
Quantity 500 g
Synonym 4-Azahepta-1,6-diyne; N,N-Di(prop-2-ynyl)amine; N-(2-Propyn-1-yl)-2-propyn-1-amine; N-(2-Propynyl)-2-propyn-1-amine; Di-2-propynylamine; NSC 80652
Molecular Weight (g/mol) 93.13
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.