accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NNOHCl

CAS RN 69221-14-3

IUPAC Name: 1-benzyl-5-ethenyl-2-[hydroxy(quinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium chloride
Synonyms: 1-benzyl-5-ethenyl-2-[hydroxy(quinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium chloride
Molecular Weight (g/mol): 420.98
Molecular Formula: C26H29ClN2O
InChi Key: FCHYSBWCOKEPNQ-UHFFFAOYNA-M
SMILES: [Cl-].OC(C1CC2CC[N+]1(CC1=CC=CC=C1)CC2C=C)C1=C2C=CC=CC2=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results