Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylcinchoninium Chloride 99.0+%, TCI America™
SDP

Catalog No. B16895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16895G Supplier TCI America Supplier No. B16895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69221-14-3
Molecular Formula C26H29ClN2O
Molecular Weight (g/mol) 420.981
MDL Number MFCD00082423
InChI Key FCHYSBWCOKEPNQ-QOVGZWAVSA-M
Synonym BCNC
PubChem CID 131857705
IUPAC Name (S)-[(1S,2R,4S,5R)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;chloride
SMILES C=CC1C[N+]2(CCC1CC2C(C3=CC=NC4=CC=CC=C34)O)CC5=CC=CC=C5.[Cl-]

Specifications

Color White
Quantity 5 g
Formula Weight 420.98
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylcinchoninium Chloride [Chiral Phase-Transfer Catalyst]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.