accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3NH3C

CAS RN 69227-51-6

IUPAC Name: 1-ethyl-1-methylpyrrolidin-1-ium bromide
Synonyms: 1-ethyl-1-methylpyrrolidin-1-ium bromide
Molecular Weight (g/mol): 194.12
Molecular Formula: C7H16BrN
InChi Key: KHJQQUGSPDBDRM-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1(C)CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)