Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-1-Methylpyrrolidinium Bromide

Catalog No. 7131715G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-317-15G 5 g
71-317-150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 71-317-15G Supplier Sigma Aldrich Supplier No. 7131715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-1-methylpyrrolidinium bromide is an ionic liquid that can be prepared by reacting N-methylpyrrolidine with bromoethane. it is conventionally used as a bromine-sequestering agent (BSA) in zinc/bromine redox flow batteries.1-Ethyl-1-methylpyrrolidinium bromide may be used in the preparation of 1-ethyl-1-methylpyrrolidiniu mLactate, a potential solvent for the separation of hexan-1-ene from hexane. It can also be used to prepare 1-ethyl-1-methylpyrrolidinium tribromide. It can be used as a co-catalyst during the cycloaddition of CO2 with different epoxides to form cyclic carbonates in the presence of a nona-vacant Keggin-type tricarbonyl rhenium derivative catalyst.

Specifications

CAS 69227-51-6
Molecular Formula C7H16BrN
Linear Formula C7H16BrN
MDL Number MFCD03095384
Quantity 5 g
Molecular Weight (g/mol) 194.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.