accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNOONOOH2OCl

CAS RN 69231-13-6

IUPAC Name: 2,3-bis(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium hydrate chloride
Synonyms: 2,3-bis(4-nitrophenyl)-5-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium hydrate chloride
Molecular Weight (g/mol): 442.82
Molecular Formula: C19H15ClN6O5
InChi Key: XBAVDBNQELUZNQ-UHFFFAOYSA-M
SMILES: O.[Cl-].[O-][N+](=O)C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results