Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium Chloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. B1047100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1047100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1047100MG Supplier TCI America Supplier No. B1047100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69231-13-6
Molecular Formula C19H15ClN6O5
Molecular Weight (g/mol) 442.816
MDL Number MFCD00060010
InChI Key XBAVDBNQELUZNQ-UHFFFAOYSA-M
PubChem CID 87175947
IUPAC Name 2,3-bis(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride;hydrate
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=C(C=C3)[N+](=O)[O-])C4=CC=C(C=C4)[N+](=O)[O-].O.[Cl-]

Specifications

Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 424.80
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium Chloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.