accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBr

CAS RN 69249-61-2

IUPAC Name: 2-bromo-3-hexylthiophene
Synonyms: 2-bromo-3-hexylthiophene
Molecular Weight (g/mol): 247.19
Molecular Formula: C10H15BrS
InChi Key: XQJNXCHDODCAJF-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(Br)SC=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)