Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2-Bromo-3-hexylthiophene, 98%

Catalog No. AAH5624303
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AAH56243-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56243-03 Supplier Thermo Scientific™ Supplier No. H5624303

missing translation for 'validMainframeMsg'

Specifications

CAS 69249-61-2
Density 1.240
Flash Point 110°(230°F)
Molecular Formula C{1}{0}H{1}{5}BrS
Quantity 1 g
MDL Number MFCD09907959
Formula Weight 247.20
Percent Purity 98%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.