accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClClClClOOCH3

CAS RN 6928-67-2

IUPAC Name: 1,2-dipropyl 3,4,5,6-tetrachlorobenzene-1,2-dicarboxylate
Synonyms: 1,2-dipropyl 3,4,5,6-tetrachlorophthalate
Molecular Weight (g/mol): 388.06
Molecular Formula: C14H14Cl4O4
InChi Key: QJRSPJSHRYYBJL-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1C(=O)OCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results