Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropyl Tetrachlorophthalate 98.0+%, TCI America™
SDP

Catalog No. T05955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T05955G Supplier TCI America Supplier No. T05955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6928-67-2
Molecular Formula C14H14Cl4O4
Molecular Weight (g/mol) 388.062
MDL Number MFCD00059245
InChI Key QJRSPJSHRYYBJL-UHFFFAOYSA-N
Synonym Tetrachlorophthalic Acid Dipropyl Ester
PubChem CID 81346
IUPAC Name dipropyl 3,4,5,6-tetrachlorobenzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OCCC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 388.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Dipropyl Tetrachlorophthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.