accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 693-07-2

IUPAC Name: 1-chloro-2-(ethylsulfanyl)ethane
Synonyms: 2-chloroethyl ethyl sulfide
Molecular Weight (g/mol): 124.63
Molecular Formula: C4H9ClS
InChi Key: GBNVXYXIRHSYEG-UHFFFAOYSA-N
SMILES: CCSCCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)