Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Ethyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. C09385ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09385ML 5 mL
C093825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09385ML Supplier TCI America Supplier No. C09385ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 693-07-2
Molecular Formula C4H9ClS
Molecular Weight (g/mol) 124.63
MDL Number MFCD00000979
InChI Key GBNVXYXIRHSYEG-UHFFFAOYSA-N
Synonym 2-chloroethyl ethyl sulfide, 1-chloro-2-ethylthio ethane, ethane, 1-chloro-2-ethylthio, half-mustard gas, 2-ethylthioethyl chloride, 2-ethylthio chloroethane, sulfide, chloroethyl ethyl, ethyl 2-chloroethyl sulfide, h-mg, sulfide, 2-chloroethyl ethyl
PubChem CID 12733
IUPAC Name 1-chloro-2-(ethylsulfanyl)ethane
SMILES CCSCCCl

Specifications

Color Yellow
Boiling Point 157°C
Quantity 5 mL
UN Number 1992
Formula Weight 124.63
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Ethyl Sulfide
RTECSNumber WQ3250000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.