accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 693-25-4

H3CMgBr

CAS RN 693-25-4

IUPAC Name: bromo(pentyl)magnesium
Synonyms: bromo(pentyl)magnesium
Molecular Weight (g/mol): 175.35
Molecular Formula: C5H11BrMg
InChi Key: ZXWGTPYWPZGZPT-UHFFFAOYSA-M
SMILES: CCCCC[Mg]Br
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results