Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentylmagnesium Bromide Solution

Catalog No. 290998800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-099-8800ML 800 mL
29-099-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-099-8800ML Supplier Sigma Aldrich Supplier No. 290998800ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentylmagnesium bromide solution is a Grignard reagent that can be used in the preparation of: Alkanes via Ni or Cu-catalyzed cross-coupling reaction with alkyl fluorides.Hydroxypiperidinones via the addition-cyclization-deprotection process of aldimines.It can also be used as a reagent to synthesize a key intermediate by olefin alkylation in the total synthesis of alkaloid, (+)-hyperaspine , and surface alkylation of H-terminated Si (111), by Si-C bond formation.

Specifications

CAS 693-25-4
Density 0.947 g/mL at 25°C
Molecular Formula C5H11BrMg
Linear Formula C5H11MgBr
Quantity 800 mL
MDL Number MFCD00000045
Molecular Weight (g/mol) 175.35 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.